Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495309.2 | XP_003495357.1 | 1 | 1318 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |