Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501135.2 | XP_003501183.1 | 3 - 4 | 774 | 69 | |
XM_007641834.1 | XP_007640024.1 | 2 - 3 | 505 | 69 | ||
XM_007641835.1 | XP_007640025.1 | 3 - 4 | 783 | 69 | ||
GenBank mRNA | JI869455.1 | - | 3 - 4 | 495 | 69 | |
JP062235.1 | - | 3 - 4 | 495 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |