Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpihbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510445.1 | XP_003510493.1 | 6 - 7 | 776 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |