Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512952.1 | XP_003513000.1 | 4 - 5 | 576 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |