Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509281.1 | XP_003509329.1 | 3 - 4 | 173 | 66 | |
XM_007649855.1 | XP_007648045.1 | 3 - 4 | 155 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |