Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509281.1 | XP_003509329.1 | 4 - 5 | 343 | 89 | |
XM_007649855.1 | XP_007648045.1 | 4 - 5 | 325 | 89 | ||
XM_007649856.1 | XP_007648046.1 | 3 - 4 | 283 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |