Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643486.1 | XP_007641676.1 | 1 | 219 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |