Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505033.2 | XP_003505081.1 | 3 - 4 | 218 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |