Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514938.2 | XP_003514986.2 | 7 - 8 | 1034 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Probe spans exons |