Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sac3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654681.1 | XP_007652871.1 | 3 | 717 | 91 | |
GenBank mRNA | JI873434.1 | - | 2 | 493 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |