Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709429.3 | XP_005709486.2 | 2 - 3 | 285 | 71 | |
GenBank mRNA | KA307661.1 | - | 2 - 3 | 572 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |