Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699885.3 | XP_005699942.1 | 2 - 3 | 250 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |