Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058028.1 | XP_017913517.1 | 2 - 3 | 532 | 68 | |
GenBank mRNA | JO438130.2 | - | 2 - 3 | 354 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |