Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971571.2 | XP_013827025.1 | 3 - 4 | 474 | 66 | |
GenBank mRNA | JO514999.2 | - | 3 - 4 | 451 | 66 | |
JO631216.2 | - | 545 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |