Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050270.1 | XP_017905759.1 | 2 - 3 | 279 | 89 | |
XM_018050271.1 | XP_017905760.1 | 2 - 3 | 394 | 89 | ||
GenBank mRNA | KA350914.1 | - | 2 - 3 | 1181 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |