Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285744.1 | NP_001272673.1 | 5 - 6 | 658 | 126 | |
GenBank mRNA | GU985267.1 | - | 5 - 6 | 658 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |