Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285701.1 | NP_001272630.1 | 3 - 4 | 348 | 89 | |
XM_013971605.2 | XP_013827059.1 | 4 - 5 | 541 | 89 | ||
GenBank mRNA | AY466498.1 | - | 3 - 4 | 351 | 89 | |
FJ844408.1 | - | 3 - 4 | 350 | 89 | ||
HQ404383.1 | - | 3 - 4 | 347 | 89 | ||
JQ031287.1 | - | 3 - 4 | 349 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |