Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285593.1 | NP_001272522.1 | 1 - 2 | 94 | 102 | |
GenBank mRNA | JN561130.1 | - | 1 - 2 | 94 | 102 | |
JX627316.1 | - | 1 - 2 | 94 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |