Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052134.1 | XP_017907623.1 | 4 - 5 | 627 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |