Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041585.1 | XP_017897074.1 | 4 - 5 | 781 | 71 | |
GenBank mRNA | KA310371.1 | - | 1 - 2 | 1930 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |