Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059873.1 | XP_017915362.1 | 3 - 4 | 3599 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |