Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674844.3 | XP_005674901.3 | 3 - 4 | 643 | 70 | |
XM_005674845.3 | XP_005674902.3 | 3 - 4 | 321 | 70 | ||
XM_005674846.3 | XP_005674903.3 | 2 - 3 | 279 | 70 | ||
XM_005674847.3 | XP_005674904.3 | 2 - 3 | 594 | 70 | ||
GenBank mRNA | JO408603.2 | - | 1 - 2 | 145 | 70 | |
JO602579.2 | - | 2 - 3 | 557 | 70 | ||
JO616961.2 | - | 1 - 2 | 81 | 70 | ||
JT705545.1 | - | 3 - 4 | 540 | 70 | ||
JT717662.1 | - | 3 - 4 | 410 | 70 | ||
KA320642.1 | - | 263 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |