Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676905.3 | XP_005676962.2 | 4 - 5 | 753 | 76 | |
XM_005676906.3 | XP_005676963.1 | 3 - 4 | 408 | 76 | ||
GenBank mRNA | JO434016.2 | - | 4 - 5 | 612 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |