Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698968.3 | XP_005699025.2 | 3 - 4 | 535 | 91 | |
GenBank mRNA | JI861806.1 | - | 3 - 4 | 481 | 91 | |
JO578817.1 | - | 3 - 4 | 601 | 91 | ||
KA320701.1 | - | 3 - 4 | 769 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |