Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968196.2 | XP_013823650.2 | 1 - 2 | 635 | 77 | |
XM_018056541.1 | XP_017912030.1 | 1 - 2 | 635 | 77 | ||
XM_018056542.1 | XP_017912031.1 | 2 - 3 | 363 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |