Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059090.1 | XP_017914579.1 | 2 - 3 | 469 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |