Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682878.1 | XP_005682935.1 | 2 - 3 | 459 | 55 | |
GenBank mRNA | KA310342.1 | - | 6 - 7 | 816 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |