Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690895.3 | XP_005690952.1 | 3 - 4 | 1274 | 72 | |
XM_013970438.2 | XP_013825892.1 | 3 - 4 | 1258 | 72 | ||
XM_018060613.1 | XP_017916102.1 | 3 - 4 | 1118 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |