Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H9orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052630.1 | XP_017908119.1 | 7 - 8 | 835 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |