Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H1orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678339.3 | XP_005678396.2 | 5 - 6 | 866 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |