Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695646.3 | XP_005695703.2 | 1 - 2 | 221 | 63 | |
GenBank mRNA | JO537556.2 | - | 1 - 2 | 783 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |