Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056478.1 | XP_017911967.1 | 1 - 2 | 911 | 63 | |
GenBank mRNA | JT681476.1 | - | 2 - 3 | 279 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |