Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065846.1 | XP_017921335.1 | 2 | 1748 | 100 | |
GenBank mRNA | JO622653.2 | - | 1 | 1685 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |