Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062603.1 | XP_017918092.1 | 1 - 2 | 679 | 97 | |
XM_018062604.1 | XP_017918093.1 | 1 - 2 | 681 | 97 | ||
GenBank mRNA | FJ455501.1 | - | 1 - 2 | 137 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |