Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687208.3 | XP_005687265.1 | 3 - 4 | 357 | 83 | |
XM_005687209.3 | XP_005687266.1 | 3 - 4 | 249 | 83 | ||
GenBank mRNA | JO584870.2 | - | 3 - 4 | 331 | 83 | |
KA316541.1 | - | 3 - 4 | 349 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |