Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057532.1 | XP_017913021.1 | 2 - 3 | 725 | 76 | |
GenBank mRNA | KA339345.1 | - | 6 - 7 | 3018 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |