Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051196.1 | XP_017906685.1 | 5 - 6 | 526 | 77 | |
XM_018051197.1 | XP_017906686.1 | 5 - 6 | 410 | 77 | ||
XM_018051199.1 | XP_017906688.1 | 4 - 5 | 337 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |