Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047287.1 | XP_017902776.1 | 4 - 5 | 3476 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Capra hircus Probe spans exons |