Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972619.2 | XP_013828073.2 | 3 - 4 | 1049 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |