Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692705.3 | XP_005692762.2 | 4 - 5 | 2328 | 64 | |
GenBank mRNA | JP350892.2 | - | 5 - 6 | 2022 | 64 | |
JP350894.2 | - | 5 - 6 | 2037 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |