Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055744.1 | XP_017911233.1 | 8 - 9 | 2466 | 56 | |
GenBank mRNA | JO427447.1 | - | 8 - 9 | 2446 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |