Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056586.1 | XP_017912075.1 | 3 - 4 | 432 | 92 | |
GenBank mRNA | KA319327.1 | - | 3 - 4 | 296 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |