Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058029.1 | XP_017913518.1 | 3 - 4 | 427 | 98 | |
GenBank mRNA | JO622816.2 | - | 3 - 4 | 367 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |