Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038725.1 | XP_017894214.1 | 3 - 4 | 1085 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |