Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695987.2 | XP_005696044.1 | 1 - 2 | 317 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |