Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066672.1 | XP_017922161.1 | 3 - 4 | 872 | 75 | |
GenBank mRNA | JO626856.2 | - | 3 - 4 | 856 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |