Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689133.3 | XP_005689190.2 | 3 - 4 | 804 | 63 | |
GenBank mRNA | HM006915.1 | - | 1 - 2 | 135 | 63 | |
JO401515.2 | - | 2 - 3 | 794 | 63 | ||
KA343070.1 | - | 1 - 2 | 185 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |