Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051608.1 | XP_017907097.1 | 3 - 4 | 252 | 75 | |
XM_018051609.1 | XP_017907098.1 | 3 - 4 | 247 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |