Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052699.1 | XP_017908188.1 | 3 - 4 | 1120 | 88 |
Set Membership: |
![]() ![]() ![]() |