Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973971.2 | XP_013829425.2 | 2 - 3 | 754 | 74 | |
GenBank mRNA | KA314117.1 | - | 4 - 5 | 755 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |